Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098518.1 | NP_001091988.1 | 16 | 2611 | ||
NM_015234.4 | NP_056049.4 | 16 | 2551 | |||
XM_005248892.2 | XP_005248949.1 | 17 | 2731 | |||
XM_005248893.3 | XP_005248950.1 | 17 | 2687 | |||
XM_005248894.3 | XP_005248951.1 | 17 | 2731 | |||
XM_005248895.3 | XP_005248952.1 | 17 | 2731 | |||
XM_011514353.2 | XP_011512655.1 | 16 | 2611 | |||
XM_011514354.2 | XP_011512656.1 | 18 | 2754 | |||
XM_017010418.1 | XP_016865907.1 | 17 | 2687 | |||
XM_017010419.1 | XP_016865908.1 | 16 | 2567 | |||
XM_017010420.1 | XP_016865909.1 | 16 | 2611 | |||
XM_017010421.1 | XP_016865910.1 | 16 | 2567 | |||
XM_017010422.1 | XP_016865911.1 | 16 | 2527 | |||
XM_017010423.1 | XP_016865912.1 | 17 | 2634 | |||
XM_017010424.1 | XP_016865913.1 | 17 | 2634 | |||
GenBank mRNA | AB018301.2 | - | 16 | 2549 | ||
AB031421.1 | - | 15 | 2366 | |||
AK289503.1 | - | 16 | 2549 | |||
AK296948.1 | - | 6 | 934 | |||
AK300380.1 | - | 5 | 1122 | |||
AL832125.1 | - | 14 | 2092 | |||
AY140958.1 | - | 15 | 2319 | |||
AY498875.1 | - | 16 | 2410 | |||
BC050087.1 | - | 3 | 435 | |||
BC066121.1 | - | 16 | 2389 | 5298732 |