Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010870.2 | NP_001010870.1 | 1 | 4115 | ||
NM_001168359.1 | NP_001161831.1 | 1 | 4115 | |||
GenBank mRNA | AF039442.1 | - | 1 | 52 | ||
AK125838.1 | - | 1 | 1203 | |||
BX648686.1 | - | 1 | 647 | |||
CR936792.1 | - | 1 | 2465 | |||
EF185284.1 | - | 1 | 600 |