Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199159.1 | NP_001186088.1 | 7 | 1599 | ||
NM_153370.2 | NP_699201.2 | 6 | 1704 | |||
XM_005248917.2 | XP_005248974.1 | 6 | 954 | |||
XM_011514375.2 | XP_011512677.1 | 6 | 1533 | |||
XM_017010430.1 | XP_016865919.1 | 6 | 954 | |||
GenBank mRNA | AK075470.1 | - | 6 | 1399 | ||
AK124589.1 | - | 9 | 2644 | |||
AK302193.1 | - | 5 | 1207 | |||
AK303928.1 | - | 5 | 1353 | |||
AK312785.1 | - | 6 | 1397 | |||
AL050088.1 | - | 1 | 120 | |||
AY358422.1 | - | 6 | 1390 | |||
BC022399.1 | - | 6 | 1399 | 4691115 | ||
BC035634.1 | - | 6 | 1704 | 5217121 |