Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079653.1 | NP_001073121.1 | 14 | 2021 | ||
NM_152744.3 | NP_689957.3 | 40 | 5876 | |||
XM_011515188.1 | XP_011513490.1 | 34 | 4717 | |||
XM_011515189.1 | XP_011513491.1 | 34 | 4714 | |||
XM_017011837.1 | XP_016867326.1 | 34 | 4717 | |||
GenBank mRNA | AK074077.1 | - | 7 | 816 | ||
AK074083.1 | - | 25 | 3512 | |||
AL833354.1 | - | 14 | 2018 | |||
AY310398.1 | - | 40 | 5876 |