Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100600.1 | NP_001094070.1 | 3, 4 | 491 | ||
NM_001270375.1 | NP_001257304.1 | 3, 4 | 491 | |||
NM_198403.3 | NP_940685.3 | 3, 4 | 491 | |||
NR_072989.1 | 3, 4 | 491 | ||||
GenBank mRNA | AY424288.1 | - | 3, 4 | 400 |