Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THSD7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015204.2 | NP_056019.1 | 14 | 3418 | ||
XM_006715659.1 | XP_006715722.1 | 14 | 3417 | |||
XM_006715660.1 | XP_006715723.1 | 14 | 3417 | |||
XM_006715662.1 | XP_006715725.1 | 14 | 3417 | |||
XM_011515193.2 | XP_011513495.1 | 14 | 5742 | |||
XM_011515194.1 | XP_011513496.1 | 14 | 3183 | |||
GenBank mRNA | AB023177.2 | - | 13 | 2702 |