Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NYAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173564.3 | NP_775835.2 | 4 | 1771 | ||
XM_006715907.3 | XP_006715970.1 | 4 | 1826 | |||
XM_017011868.1 | XP_016867357.1 | 4 | 1826 | |||
GenBank mRNA | AK124773.1 | - | 4 | 1771 |