Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308028.1 | NP_001294957.1 | 14 | 1893 | ||
NM_001308031.1 | NP_001294960.1 | 7 | 843 | |||
NM_005246.3 | NP_005237.2 | 16 | 2279 | |||
XM_011543266.1 | XP_011541568.1 | 16 | 2132 | |||
XM_011543267.1 | XP_011541569.1 | 16 | 4016 | |||
XM_011543269.1 | XP_011541571.1 | 15 | 3870 | |||
XM_011543270.2 | XP_011541572.1 | 15 | 2047 | |||
XM_011543271.2 | XP_011541573.1 | 16 | 2132 | |||
XM_011543272.1 | XP_011541574.1 | 15 | 1928 | |||
XM_011543276.2 | XP_011541578.1 | 7 | 1050 | |||
XM_017009229.1 | XP_016864718.1 | 17 | 2279 | |||
XM_017009230.1 | XP_016864719.1 | 15 | 2047 | |||
XM_017009231.1 | XP_016864720.1 | 15 | 3870 | |||
XM_017009232.1 | XP_016864721.1 | 15 | 1920 | |||
XM_017009233.1 | XP_016864722.1 | 14 | 1863 | |||
GenBank mRNA | AK299855.1 | - | 14 | 1858 | ||
AK315234.1 | - | 14 | 1967 | |||
J03358.1 | - | 16 | 2279 | |||
JQ412173.1 | - | 7 | 843 |