Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000034.3 | NP_000025.1 | 8, 9 | 1401 | ||
NM_001127617.2 | NP_001121089.1 | 3, 4 | 623 | |||
NM_001243177.1 | NP_001230106.1 | 4, 5 | 744 | |||
NM_184041.2 | NP_908930.1 | 3, 4 | 590 | |||
NM_184043.2 | NP_908932.1 | 3, 4 | 562 | |||
XM_011545768.2 | XP_011544070.1 | 5, 6 | 1264 | |||
GenBank mRNA | AK026577.1 | - | 3, 4 | 440 | ||
AK098778.1 | - | 6, 7 | 1442 | |||
AK301993.1 | - | 4, 5 | 556 | |||
BC004333.1 | - | 8, 9 | 1321 | 3630974 | ||
BC010660.1 | - | 3, 4 | 508 | 3869603 | ||
BC012880.1 | - | 3, 4 | 436 | 3459009 | ||
BC013614.1 | - | 3, 4 | 519 | 3880857 | ||
BC015888.2 | - | 3, 4 | 432 | 3911623 | ||
BC016800.1 | - | 4, 5 | 460 | 4072487 | ||
BQ719865.1 | - | 3, 4 | 551 | |||
BX647566.1 | - | 3, 4 | 412 | |||
CR536528.1 | - | 2, 3 | 314 | |||
CR541880.1 | - | 2, 3 | 314 | |||
DA893541.1 | - | 3, 4 | 470 | |||
FJ474908.1 | - | 4, 5 | 481 | |||
M11560.1 | - | 4, 5 | 481 |