Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004439.1 | NP_001004439.1 | 24 | 3044 | ||
XM_005254228.3 | XP_005254285.1 | 22 | 3266 | |||
XM_011521363.2 | XP_011519665.1 | 22 | 2996 | |||
GenBank mRNA | AF109681.1 | - | 24 | 3029 | ||
AF111799.1 | - | 16 | 2009 | |||
AF137378.2 | - | 24 | 3047 | |||
AK292445.1 | - | 24 | 3053 | |||
AL359064.1 | - | 2 | 100 |