Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317960.1 | NP_001304889.1 | 24, 25 | 2524 | ||
NM_012255.4 | NP_036387.2 | 24, 25 | 2364 | |||
XM_017027723.1 | XP_016883212.1 | 24, 25 | 2317 | |||
GenBank mRNA | AF064257.1 | - | 24, 25 | 2317 | ||
AF152169.1 | - | 24, 25 | 2267 | |||
AK000084.1 | - | 13, 14 | 1179 | |||
AK001573.1 | - | 24, 25 | 2334 | |||
AK302846.1 | - | 24, 25 | 2228 | |||
AK303312.1 | - | 23, 24 | 2175 | |||
AK314602.1 | - | 24, 25 | 2303 | |||
AL136841.1 | - | 24, 25 | 2290 | |||
AY382900.1 | - | 23, 24 | 2179 | |||
BC006417.1 | - | 12, 13 | 1139 |