Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014280.2 | NP_055095.2 | 2 | 188 | ||
GenBank mRNA | AF083190.1 | - | 2 | 195 | ||
AK300797.1 | - | 2 | 177 | |||
BC033159.1 | - | 2 | 173 | 4330676 | ||
BX538122.1 | - | 3 | 305 | |||
CR456900.1 | - | 2 | 188 | |||
DB294321.1 | - | 2 | 188 |