Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042486.3 | NP_001035951.1 | 7 | 1388 | ||
NM_014902.5 | NP_055717.2 | 13 | 3424 | |||
NM_183006.3 | NP_892118.1 | 7 | 2109 | |||
XM_005260332.4 | XP_005260389.1 | 7 | 1326 | |||
XM_005260333.4 | XP_005260390.1 | 8 | 1415 | |||
XM_011528688.2 | XP_011526990.1 | 12 | 3199 | |||
XM_011528691.2 | XP_011526993.1 | 8 | 2248 | |||
XM_011528692.2 | XP_011526994.1 | 8 | 1438 | |||
XM_017027725.1 | XP_016883214.1 | 7 | 2136 | |||
XM_017027726.1 | XP_016883215.1 | 7 | 1345 | |||
XM_017027727.1 | XP_016883216.1 | 6 | 1275 | |||
GenBank mRNA | AB023181.1 | - | 13 | 3366 | ||
AK001681.1 | - | 7 | 1335 | |||
AK055152.1 | - | 6 | 2436 | |||
AK125383.1 | - | 7 | 2157 | |||
BC023214.1 | - | 7 | 1506 | |||
BC108706.1 | - | 7 | 2098 | |||
BC153874.1 | - | 7 | 2109 |