Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042486.3 | NP_001035951.1 | 4 | 619 | ||
NM_014902.5 | NP_055717.2 | 10 | 2655 | |||
NM_183006.3 | NP_892118.1 | 4 | 1340 | |||
XM_005260332.4 | XP_005260389.1 | 4 | 557 | |||
XM_005260333.4 | XP_005260390.1 | 5 | 646 | |||
XM_011528688.2 | XP_011526990.1 | 9 | 2430 | |||
XM_011528689.2 | XP_011526991.1 | 9 | 2437 | |||
XM_011528691.2 | XP_011526993.1 | 5 | 1479 | |||
XM_011528692.2 | XP_011526994.1 | 5 | 669 | |||
XM_017027725.1 | XP_016883214.1 | 4 | 1367 | |||
XM_017027726.1 | XP_016883215.1 | 4 | 576 | |||
XM_017027727.1 | XP_016883216.1 | 3 | 506 | |||
GenBank mRNA | AB023181.1 | - | 10 | 2597 | ||
AK055152.1 | - | 3 | 1667 | |||
AK125383.1 | - | 4 | 1317 | |||
BC023214.1 | - | 4 | 737 | |||
BC108706.1 | - | 4 | 1329 | |||
BC153874.1 | - | 4 | 1340 | |||
CN419826.1 | - | 3 | 483 |