Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178137.1 | NP_001171608.1 | 9 | 1813 | ||
NM_014912.4 | NP_055727.3 | 9 | 1937 | |||
XM_005269630.4 | XP_005269687.1 | 9 | 1895 | |||
XM_006717714.3 | XP_006717777.1 | 10 | 1906 | |||
XM_006717715.3 | XP_006717778.1 | 10 | 1988 | |||
XM_011539514.2 | XP_011537816.1 | 9 | 1964 | |||
XM_011539515.2 | XP_011537817.1 | 10 | 1919 | |||
XM_011539517.2 | XP_011537819.1 | 8 | 1838 | |||
XM_011539518.2 | XP_011537820.1 | 10 | 1988 | |||
XM_011539519.2 | XP_011537821.1 | 7 | 1706 | |||
XM_011539520.2 | XP_011537822.1 | 7 | 1637 | |||
XM_017015927.1 | XP_016871416.1 | 6 | 701 | |||
GenBank mRNA | AB023157.1 | - | 9 | 1780 | ||
AK127060.1 | - | 8 | 885 | |||
AK302576.1 | - | 9 | 1882 | |||
BC036444.2 | - | 9 | 1903 | 4838004 |