Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135208.1 | NP_001128680.1 | 4, 5 | 436 | ||
NM_004470.3 | NP_004461.2 | 4, 5 | 521 | |||
NM_057092.2 | NP_476433.1 | 4, 5 | 427 | |||
XM_005273848.3 | XP_005273905.1 | 4, 5 | 423 | |||
GenBank mRNA | AW450265.1 | - | 2, 3 | 223 | ||
BC003384.1 | - | 4, 5 | 427 | 3445148 | ||
BC091475.1 | - | 4, 5 | 373 | 30349098 | ||
BT009861.1 | - | 3, 4 | 325 | |||
BU682774.1 | - | 2, 3 | 212 | |||
CR407636.1 | - | 3, 4 | 325 | |||
M65128.1 | - | 4, 5 | 359 | |||
M75099.1 | - | 4, 5 | 355 |