Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002013.3 | NP_002004.1 | 3 | 316 | ||
XM_011536565.2 | XP_011534867.1 | 3 | 2337 | |||
XM_017021092.1 | XP_016876581.1 | 3 | 265 | |||
XM_017021093.1 | XP_016876582.1 | 3 | 2337 | |||
GenBank mRNA | AK311915.1 | - | 3 | 254 | ||
BC016288.1 | - | 4 | 654 | 3919642 | ||
BC020809.1 | - | 3 | 254 | 4292678 | ||
BI598048.1 | - | 3 | 321 | |||
BT006904.1 | - | 3 | 243 | |||
M96256.1 | - | 3 | 266 |