Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SACM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319073.1 | NP_001306002.1 | 4, 5 | 461 | ||
NM_014016.4 | NP_054735.3 | 3, 4 | 421 | |||
GenBank mRNA | AB020658.1 | - | 3, 4 | 265 | ||
AK291142.1 | - | 3, 4 | 305 | |||
AK301248.1 | - | 4, 5 | 301 | |||
AL136831.1 | - | 3, 4 | 211 | |||
BC016559.1 | - | 3, 4 | 217 | 3049075 | ||
CR749564.1 | - | 3, 4 | 1367 | |||
DC317560.1 | - | 3, 4 | 400 |