Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012309.4 | NP_036441.2 | 22 | 2582 | ||
NM_133266.4 | NP_573573.2 | 8 | 779 | |||
NR_110766.1 | 8 | 779 | ||||
XM_005277932.3 | XP_005277989.1 | 13 | 1587 | |||
XM_017017387.1 | XP_016872876.1 | 23 | 2687 | |||
XM_017017388.1 | XP_016872877.1 | 23 | 2709 | |||
XM_017017389.1 | XP_016872878.1 | 23 | 2882 | |||
XM_017017390.1 | XP_016872879.1 | 10 | 3820 | |||
GenBank mRNA | AB028945.1 | - | 5 | 316 | ||
AB208025.1 | - | 22 | 2548 | |||
AB208026.1 | - | 13 | 1370 | |||
AB208027.1 | - | 8 | 757 | |||
AF141901.1 | - | 8 | 671 | |||
BC093885.1 | - | 8 | 779 | |||
BC112097.1 | - | 8 | 779 | |||
BC114484.1 | - | 8 | 701 | |||
BC143389.1 | - | 6 | 749 |