Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SCMH1 SIRNA Assays ›
Gene Symbol:
SCMH1
Gene Name:
sex comb on midleg homolog 1 (Drosophila)
Gene Aliases:
Scml3
Chromosome Location:
Chr.1: 41027199 - 41242155 on Build GRCh38
UniGene ID:
Hs.571874
Species:
Human
siRNA ID
HSS177056
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SCMH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031694.2
NP_001026864.1
8, 9
1087
NM_001172218.1
NP_001165689.1
9, 10
1159
NM_001172219.1
NP_001165690.1
8, 9
950
NM_001172220.1
NP_001165691.1
7, 8
879
NM_001172221.1
NP_001165692.1
7, 8
974
NM_012236.3
NP_036368.1
8, 9
1088
NR_135092.1
8, 9
1087
XM_006710462.2
XP_006710525.1
8, 9
950
XM_006710464.1
XP_006710527.1
7, 8
899
XM_011541032.2
XP_011539334.1
8, 9
888
XM_011541033.2
XP_011539335.1
10, 11
1305
XM_011541034.1
XP_011539336.1
8, 9
950
XM_011541035.2
XP_011539337.1
8, 9
1098
XM_011541036.2
XP_011539338.1
9, 10
1161
XM_011541040.2
XP_011539342.1
8, 9
950
XM_011541043.2
XP_011539345.1
10, 11
1401
XM_011541044.1
XP_011539346.1
3, 4
517
XM_011541045.2
XP_011539347.2
9, 10
1174
XM_017000698.1
XP_016856187.1
5, 6
703
XM_017000699.1
XP_016856188.1
7, 8
889
XM_017000700.1
XP_016856189.1
8, 9
1099
XM_017000702.1
XP_016856191.1
8, 9
964
XM_017000703.1
XP_016856192.1
8, 9
1102
XM_017000709.1
XP_016856198.1
7, 8
926
GenBank mRNA
AF149045.1
-
8, 9
1061
AF149046.1
-
9, 10
1132
AK023124.1
-
8, 9
917
AK301497.1
-
6, 7
929
BC009752.2
-
7, 8
1029
3010273
BC021252.2
-
7, 8
987
3355251
BX640721.1
-
7, 8
865
CR457161.1
-
3, 4
535
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
spermatogenesis
anatomical structure morphogenesis
anterior/posterior pattern specification
gene silencing
protein sumoylation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0