Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014991.4 | NP_055806.2 | 68 | 10879 | ||
XM_005262858.4 | XP_005262915.1 | 70 | 10920 | |||
XM_011531757.2 | XP_011530059.1 | 69 | 10831 | |||
XM_011531759.2 | XP_011530061.1 | 68 | 10793 | |||
XM_011531760.2 | XP_011530062.1 | 68 | 10731 | |||
XM_011531761.2 | XP_011530063.1 | 69 | 10786 | |||
XM_011531762.2 | XP_011530064.1 | 68 | 10780 | |||
XM_011531763.2 | XP_011530065.1 | 67 | 10677 | |||
XM_011531764.2 | XP_011530066.1 | 68 | 10747 | |||
XM_011531765.2 | XP_011530067.1 | 65 | 10482 | |||
XM_017007906.1 | XP_016863395.1 | 71 | 11048 | |||
XM_017007907.1 | XP_016863396.1 | 71 | 11045 | |||
XM_017007908.1 | XP_016863397.1 | 70 | 10977 | |||
XM_017007909.1 | XP_016863398.1 | 56 | 8673 | |||
GenBank mRNA | AB023210.2 | - | 33 | 4562 | ||
AF538685.1 | - | 67 | 10771 | |||
AK055806.1 | - | 19 | 2241 | |||
AK092394.1 | - | 8 | 1258 | |||
BC013377.2 | - | 5 | 1223 | |||
BC015214.1 | - | 8 | 1205 | |||
BC065502.1 | - | 15 | 2229 | |||
BC119633.2 | - | 5 | 926 | |||
BC148294.1 | - | 5 | 926 | |||
BC148298.2 | - | 5 | 926 | |||
BC150246.1 | - | 33 | 4583 | |||
BX648077.1 | - | 25 | 3444 | |||
BX648153.1 | - | 28 | 3772 |