Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015021.1 | NP_055836.1 | 8 | 6774 | ||
XM_005248697.4 | XP_005248754.1 | 8 | 6588 | |||
XM_011535625.2 | XP_011533927.1 | 7 | 6594 | |||
XM_017010575.1 | XP_016866064.1 | 9 | 9965 | |||
XM_017010576.1 | XP_016866065.1 | 10 | 7491 | |||
XM_017010577.1 | XP_016866066.1 | 8 | 9820 | |||
XM_017010578.1 | XP_016866067.1 | 10 | 12473 | |||
XM_017010579.1 | XP_016866068.1 | 10 | 9499 | |||
XM_017010580.1 | XP_016866069.1 | 10 | 6920 | |||
XM_017010581.1 | XP_016866070.1 | 9 | 6906 | |||
XM_017010582.1 | XP_016866071.1 | 8 | 9616 | |||
XM_017010583.1 | XP_016866072.1 | 9 | 7213 | |||
XM_017010584.1 | XP_016866073.1 | 8 | 11589 | |||
GenBank mRNA | AB011102.1 | - | 1 | 3254 | ||
AK023712.1 | - | 1 | 1058 | |||
AY684300.1 | - | 1 | 418 | |||
BC063594.1 | - | 1 | 461 | |||
BX537746.1 | - | 8 | 6748 |