Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178140.3 | NP_835260.2 | 20 | 8042 | ||
XM_005248269.4 | XP_005248326.1 | 20 | 8064 | |||
XM_005248271.1 | XP_005248328.1 | 19 | 7416 | |||
XM_005248272.3 | XP_005248329.1 | 19 | 7418 | |||
XM_006714460.2 | XP_006714523.1 | 19 | 7215 | |||
XM_011513992.2 | XP_011512294.1 | 21 | 8426 | |||
XM_011513993.2 | XP_011512295.1 | 20 | 8344 | |||
XM_011513994.2 | XP_011512296.1 | 21 | 8391 | |||
XM_011513995.2 | XP_011512297.1 | 21 | 8425 | |||
XM_011513996.2 | XP_011512298.1 | 19 | 8150 | |||
XM_017009245.1 | XP_016864734.1 | 17 | 7140 | |||
XM_017009246.1 | XP_016864735.1 | 21 | 8342 | |||
GenBank mRNA | AB002298.2 | - | 20 | 8042 | ||
AF338650.1 | - | 15 | 7060 |