Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161560.2 | NP_001155032.1 | 29 | 3922 | ||
NM_001161561.2 | NP_001155033.1 | 29 | 3859 | |||
NM_001161562.2 | NP_001155034.1 | 28 | 3835 | |||
NM_001161563.2 | NP_001155035.1 | 29 | 3781 | |||
NM_001161564.2 | NP_001155036.1 | 28 | 3757 | |||
NM_001161565.2 | NP_001155037.1 | 28 | 3694 | |||
NM_001161566.2 | NP_001155038.1 | 27 | 3670 | |||
NM_015028.3 | NP_055843.1 | 30 | 3946 | |||
GenBank mRNA | AB011123.2 | - | 30 | 3867 | ||
AF172264.1 | - | 30 | 3601 | |||
AF172265.1 | - | 29 | 3514 | |||
AF172266.1 | - | 29 | 3436 | |||
AF172267.1 | - | 29 | 3577 | |||
AF172268.1 | - | 28 | 3349 | |||
AF172269.1 | - | 28 | 3490 | |||
AF172270.1 | - | 28 | 3412 | |||
AF172271.1 | - | 27 | 3325 | |||
AK026470.1 | - | 4 | 720 | |||
AK291056.1 | - | 28 | 3694 | |||
BC019081.2 | - | 9 | 1065 | |||
BC055427.1 | - | 9 | 1634 | |||
BC150256.1 | - | 30 | 3867 | 9007184 |