Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015043.3 | NP_055858.2 | 21 | 3597 | ||
NM_198868.2 | NP_942568.2 | 22 | 3648 | |||
GenBank mRNA | AB014576.1 | - | 22 | 3648 | ||
AB449893.1 | - | 22 | 3611 | |||
AK001196.1 | - | 8 | 1210 | |||
AK001473.1 | - | 3 | 672 | |||
AK026096.1 | - | 1 | 159 | |||
AK074791.1 | - | 11 | 1679 | |||
AK123874.1 | - | 4 | 2064 | |||
AK223509.1 | - | 7 | 1148 | |||
BC000701.2 | - | 8 | 1231 | |||
BC008919.2 | - | 10 | 1579 | |||
BC080659.1 | - | 10 | 1658 | |||
BC110400.1 | - | 12 | 1798 | |||
BC129994.1 | - | 21 | 3598 | 40129194 | ||
BX641107.1 | - | 2 | 2314 |