Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015050.2 | NP_055865.1 | 19 | 2269 | ||
XM_005248955.3 | XP_005249012.1 | 19 | 2199 | |||
GenBank mRNA | AK292564.1 | - | 19 | 2123 | ||
BC010731.2 | - | 4 | 396 | |||
BC031890.1 | - | 19 | 2163 | 4944457 | ||
D43949.2 | - | 19 | 2159 |