Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015056.2 | NP_055871.1 | 1, 2 | 252 | ||
GenBank mRNA | AK056263.1 | - | 1, 2 | 215 | ||
BC014005.2 | - | 1, 2 | 224 | |||
BC028386.2 | - | 1, 2 | 257 | 4821672 | ||
D80001.1 | - | 1, 2 | 204 |