Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009814.1 | NP_001009814.1 | 16 | 1965 | ||
NM_015058.1 | NP_055873.1 | 16 | 1965 | |||
XM_006719791.3 | XP_006719854.1 | 8 | 921 | |||
XM_011535007.2 | XP_011533309.1 | 16 | 1991 | |||
XM_017020469.1 | XP_016875958.1 | 18 | 2024 | |||
XM_017020470.1 | XP_016875959.1 | 16 | 1991 | |||
XM_017020471.1 | XP_016875960.1 | 16 | 1991 | |||
XM_017020474.1 | XP_016875963.1 | 16 | 1991 | |||
GenBank mRNA | AB011136.1 | - | 5 | 505 | ||
AK095155.1 | - | 16 | 1839 | |||
AL834181.1 | - | 9 | 1014 | |||
BC053674.1 | - | 16 | 1965 | 6171968 | ||
BC131802.1 | - | 17 | 1798 |