Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040152.1 | NP_001035242.1 | 2 | 511 | ||
NM_001172437.2 | NP_001165908.1 | 2 | 522 | |||
NM_001172438.2 | NP_001165909.1 | 2 | 522 | |||
NM_001184961.1 | NP_001171890.1 | 2 | 511 | |||
NM_001184962.1 | NP_001171891.1 | 2 | 511 | |||
NM_015068.3 | NP_055883.2 | 2 | 511 | |||
GenBank mRNA | AB028974.1 | - | 1 | 84 | ||
AB049150.1 | - | 2 | 290 | |||
AB049834.1 | - | 1 | 149 | |||
AF216076.1 | - | 1 | 42 | |||
AK299837.1 | - | 477 | ||||
BC050659.2 | - | 2 | 443 | 6197322 | ||
BP250746.1 | - | 3 | 402 |