Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002015.3 | NP_002006.2 | 2 | 1875 | ||
XM_011535008.2 | XP_011533310.1 | 2 | 2028 | |||
XM_011535010.2 | XP_011533312.1 | 2 | 4540 | |||
GenBank mRNA | AF032885.1 | - | 2 | 1875 | ||
AK310511.1 | - | 2 | 967 | |||
BC021981.2 | - | 2 | 1803 | 2959021 | ||
BC070065.1 | - | 2 | 1866 | 30345006 | ||
BT007455.1 | - | 2 | 1490 | |||
BX648278.1 | - | 2 | 1078 | |||
U02310.1 | - | 2 | 1496 |