Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015070.4 | NP_055885.3 | 3 | 282 | ||
XM_005266301.3 | XP_005266358.1 | 3 | 576 | |||
XM_005266302.3 | XP_005266359.1 | 3 | 276 | |||
XM_005266303.3 | XP_005266360.1 | 4 | 601 | |||
XM_005266304.3 | XP_005266361.1 | 3 | 558 | |||
XM_005266305.3 | XP_005266362.1 | 4 | 594 | |||
XM_005266306.3 | XP_005266363.1 | 3 | 272 | |||
XM_005266307.3 | XP_005266364.1 | 3 | 576 | |||
XM_005266308.3 | XP_005266365.1 | 3 | 576 | |||
XM_005266309.3 | XP_005266366.1 | 3 | 276 | |||
XM_005266310.3 | XP_005266367.1 | 3 | 558 | |||
XM_005266311.3 | XP_005266368.1 | 3 | 576 | |||
XM_005266312.1 | XP_005266369.1 | 3 | 576 | |||
XM_017020475.1 | XP_016875964.1 | 4 | 364 | |||
XM_017020476.1 | XP_016875965.1 | 4 | 361 | |||
XM_017020477.1 | XP_016875966.1 | 4 | 601 | |||
XM_017020478.1 | XP_016875967.1 | 4 | 594 | |||
XM_017020479.1 | XP_016875968.1 | 4 | 364 | |||
XM_017020480.1 | XP_016875969.1 | 3 | 272 | |||
XM_017020481.1 | XP_016875970.1 | 4 | 601 | |||
XM_017020482.1 | XP_016875971.1 | 3 | 276 | |||
GenBank mRNA | AK056422.1 | - | 3 | 560 | ||
AY283618.1 | - | 3 | 280 | |||
BC019000.2 | - | 3 | 523 |