Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQSEC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111125.2 | NP_001104595.1 | 6 | 2514 | ||
NM_015075.1 | NP_055890.1 | 6 | 1791 | |||
XM_006724579.2 | XP_006724642.1 | 7 | 2636 | |||
XM_006724580.3 | XP_006724643.1 | 6 | 1804 | |||
XM_006724581.3 | XP_006724644.1 | 7 | 2636 | |||
XM_006724582.3 | XP_006724645.1 | 7 | 2636 | |||
XM_006724583.3 | XP_006724646.1 | 7 | 2636 | |||
XM_006724584.2 | XP_006724647.1 | 7 | 2636 | |||
XM_011530773.2 | XP_011529075.1 | 5 | 1708 | |||
XM_011530774.2 | XP_011529076.1 | 7 | 2636 | |||
XM_011530776.1 | XP_011529078.1 | 7 | 2636 | |||
XM_011530777.1 | XP_011529079.1 | 7 | 2636 | |||
XM_017029359.1 | XP_016884848.1 | 5 | 2510 | |||
XM_017029360.1 | XP_016884849.1 | 6 | 1922 | |||
GenBank mRNA | AB011094.1 | - | 6 | 2514 | ||
AK095232.1 | - | 6 | 1791 |