Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001455.3 | NP_001446.1 | 2 | 1578 | ||
NM_201559.2 | NP_963853.1 | 3 | 1551 | |||
XM_005266867.4 | XP_005266924.1 | 3 | 799 | |||
XM_005266868.3 | XP_005266925.1 | 2 | 823 | |||
XM_011535626.1 | XP_011533928.1 | 2 | 749 | |||
XM_011535628.2 | XP_011533930.1 | 2 | 5107 | |||
XM_011535629.2 | XP_011533931.1 | 2 | 772 | |||
XM_017010585.1 | XP_016866074.1 | 3 | 2484 | |||
XM_017010586.1 | XP_016866075.1 | 2 | 2367 | |||
GenBank mRNA | AF032886.1 | - | 2 | 2159 | ||
AK092357.1 | - | 3 | 939 | |||
AK122861.1 | - | 3 | 1134 | |||
AK301304.1 | - | 2 | 740 | |||
AK303933.1 | - | 2 | 724 | |||
AK311587.1 | - | 2 | 838 | |||
BC020227.1 | - | 3 | 1539 | 4111403 | ||
BC021224.2 | - | 3 | 1539 | 4137370 | ||
BC068552.1 | - | 2 | 1578 | 30347471 | ||
CR749261.1 | - | 2 | 808 |