Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015089.3 | NP_055904.1 | 33 | 6638 | ||
XM_011514422.1 | XP_011512724.1 | 32 | 6794 | |||
XM_011514423.2 | XP_011512725.1 | 33 | 6896 | |||
XM_011514424.1 | XP_011512726.1 | 32 | 6713 | |||
XM_011514425.1 | XP_011512727.1 | 30 | 6464 | |||
XM_011514426.1 | XP_011512728.1 | 32 | 6794 | |||
XM_011514427.1 | XP_011512729.1 | 28 | 5413 | |||
XM_011514429.1 | XP_011512731.1 | 25 | 4618 | |||
XM_017010589.1 | XP_016866078.1 | 32 | 7505 | |||
XM_017010590.1 | XP_016866079.1 | 26 | 4967 | |||
GenBank mRNA | AB014608.3 | - | 25 | 4272 | ||
AJ318215.1 | - | 32 | 6563 | |||
AY145132.1 | - | 32 | 6563 | |||
BC002879.2 | - | 10 | 1904 | |||
CR749511.1 | - | 32 | 7017 | |||
CR749841.1 | - | 33 | 6553 |