Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015123.2 | NP_055938.1 | 6 | 853 | ||
XM_005264722.1 | XP_005264779.1 | 6 | 530 | |||
XM_017005989.1 | XP_016861478.1 | 6 | 637 | |||
XM_017005990.1 | XP_016861479.1 | 6 | 530 | |||
XM_017005991.1 | XP_016861480.1 | 7 | 690 | |||
XM_017005992.1 | XP_016861481.1 | 6 | 470 | |||
XM_017005993.1 | XP_016861482.1 | 6 | 461 | |||
XM_017005994.1 | XP_016861483.1 | 6 | 2303 | |||
GenBank mRNA | AB023230.1 | - | 6 | 622 | ||
AK000244.1 | - | 6 | 434 | |||
AK295045.1 | - | 1 | 95 | |||
DC340455.1 | - | 1 | 64 |