Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015134.3 | NP_055949.2 | 21, 22 | 3036 | ||
NM_201274.3 | NP_958431.2 | 21, 22 | 3036 | |||
XM_005256563.4 | XP_005256620.1 | 22, 23 | 7148 | |||
XM_005256564.4 | XP_005256621.3 | 20, 21 | 3227 | |||
XM_011523761.2 | XP_011522063.1 | 22, 23 | 7519 | |||
XM_011523762.2 | XP_011522064.1 | 22, 23 | 7502 | |||
XM_011523763.2 | XP_011522065.1 | 21, 22 | 7388 | |||
XM_011523764.2 | XP_011522066.1 | 21, 22 | 7382 | |||
XM_011523765.2 | XP_011522067.1 | 21, 22 | 7382 | |||
XM_011523766.2 | XP_011522068.2 | 21, 22 | 3695 | |||
XM_011523767.2 | XP_011522069.2 | 21, 22 | 3695 | |||
XM_017024393.1 | XP_016879882.1 | 20, 21 | 3575 | |||
GenBank mRNA | AB020671.2 | - | 7, 8 | 4040 | ||
AB098507.1 | - | 21, 22 | 3000 | |||
AB189741.1 | - | 21, 22 | 2941 | |||
AB362572.1 | - | 4, 5 | 416 | |||
AB362573.1 | - | 4, 5 | 416 | |||
AK025229.1 | - | 11, 12 | 1701 | |||
AK025604.1 | - | 7, 8 | 3143 | |||
AK092287.1 | - | 16, 17 | 2414 | |||
AL834513.1 | - | 20, 21 | 2833 | |||
AY296247.1 | - | 21, 22 | 2944 | |||
BC009982.2 | - | 16, 17 | 2359 | |||
BC012426.1 | - | 3, 4 | 285 | |||
BC021058.1 | - | 8, 9 | 944 | |||
BC021070.1 | - | 8, 9 | 944 | |||
BC075847.1 | - | 16, 17 | 2370 | |||
BC105987.1 | - | 13, 14 | 1820 | |||
BC136479.1 | - | 21, 22 | 3024 | 9020468 | ||
D23673.1 | - | 7, 8 | 913 |