Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM175B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032182.3 | NP_115558.3 | 2 | 192 | ||
XM_011539554.1 | XP_011537856.1 | 2 | 163 | |||
XM_017015986.1 | XP_016871475.1 | 2 | 174 | |||
GenBank mRNA | AK296677.1 | - | 2 | 164 | ||
AL537809.3 | - | 2 | 144 | |||
BC008999.1 | - | 2 | 154 | 2900632 | ||
D63877.1 | - | 2 | 161 |