Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DIP2A SIRNA Assays ›
Gene Symbol:
DIP2A
Gene Name:
disco interacting protein 2 homolog A
Gene Aliases:
C21orf106, DIP2
Chromosome Location:
Chr.21: 46458942 - 46570900 on Build GRCh38
UniGene ID:
Hs.189585
Species:
Human
siRNA ID
HSS177233
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DIP2A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146115.1
NP_001139587.1
18
2334
NM_001146116.1
NP_001139588.1
19
2451
NM_015151.3
NP_055966.2
19
2463
NM_206889.2
NP_996772.1
19
2463
NM_206890.2
NP_996773.1
19
2463
NM_206891.2
NP_996774.1
19
2463
XM_011529492.2
XP_011527794.1
20
2578
XM_011529501.1
XP_011527803.1
20
2578
XM_011529502.1
XP_011527804.1
20
2578
XM_017028291.1
XP_016883780.1
20
2578
XM_017028292.1
XP_016883781.1
20
2578
XM_017028293.1
XP_016883782.1
20
2575
XM_017028294.1
XP_016883783.1
19
2457
XM_017028295.1
XP_016883784.1
19
2454
XM_017028296.1
XP_016883785.1
19
2454
XM_017028297.1
XP_016883786.1
19
2446
XM_017028298.1
XP_016883787.1
18
2325
XM_017028299.1
XP_016883788.1
19
2323
XM_017028300.1
XP_016883789.1
20
2578
XM_017028301.1
XP_016883790.1
20
2578
XM_017028302.1
XP_016883791.1
18
2218
XM_017028303.1
XP_016883792.1
20
2578
XM_017028304.1
XP_016883793.1
20
2578
XM_017028305.1
XP_016883794.1
20
2578
GenBank mRNA
AB273729.1
-
19
2301
AF490768.1
-
19
2356
AK056738.1
-
19
2355
AK091790.1
-
3
1501
AK098236.1
-
14
1606
AK303351.1
-
17
2278
BC033718.1
-
18
2317
4708615
BC038443.1
-
19
2462
5217858
BQ017399.1
-
3
421
CK821329.1
-
2
461
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
metabolic process
negative regulation of gene expression
regulation of apoptotic process
catalytic activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0