Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015156.3 | NP_055971.2 | 11 | 1575 | ||
GenBank mRNA | AF155595.1 | - | 11 | 1338 | ||
AK026612.1 | - | 1 | 115 | |||
D31888.1 | - | 11 | 1080 |