Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014614.2 | NP_055429.2 | 25 | 2874 | ||
XM_006711969.1 | XP_006712032.1 | 24 | 2451 | |||
XM_011532705.2 | XP_011531007.1 | 26 | 2898 | |||
XM_011532706.2 | XP_011531008.1 | 26 | 2898 | |||
XM_011532707.2 | XP_011531009.1 | 25 | 2832 | |||
XM_011532709.2 | XP_011531011.1 | 26 | 2898 | |||
GenBank mRNA | AB621805.1 | - | 24 | 2991 | ||
AY894754.1 | - | 24 | 2723 | |||
AY894755.1 | - | 6 | 562 | |||
BC112169.1 | - | 24 | 2510 | 8327637 | ||
BC113668.1 | - | 24 | 2510 | 8322720 | ||
BC143739.1 | - | 24 | 2510 | |||
D38521.1 | - | 25 | 2684 |