Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313858.1 | NP_001300787.1 | 3, 4 | 422 | ||
NM_015161.2 | NP_055976.1 | 3, 4 | 498 | |||
GenBank mRNA | AK300825.1 | - | 4, 5 | 306 | ||
AK300869.1 | - | 3, 4 | 367 | |||
AK301636.1 | - | 3, 4 | 422 | |||
AK308092.1 | - | 4, 5 | 282 | |||
BC010281.1 | - | 3, 4 | 355 | 3048642 | ||
BX648446.1 | - | 4, 5 | 285 | |||
D31885.1 | - | 3, 4 | 355 |