Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015166.3 | NP_055981.1 | 5 | 978 | ||
NM_139202.2 | NP_631941.1 | 5 | 617 | |||
XM_011530678.1 | XP_011528980.1 | 5 | 739 | |||
XM_017028671.1 | XP_016884160.1 | 4 | 406 | |||
GenBank mRNA | AF319633.1 | - | 5 | 540 | ||
AK124264.1 | - | 5 | 979 | |||
AK295106.1 | - | 4 | 239 | |||
AK299841.1 | - | 4 | 386 | |||
BC028425.2 | - | 5 | 577 | 4817175 | ||
BC070042.1 | - | 5 | 575 | 5285102 | ||
CR456460.1 | - | 4 | 396 | |||
D25217.3 | - | 5 | 654 |