Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253912.1 | NP_001240841.1 | 2 | 699 | ||
NM_015173.3 | NP_055988.2 | 2 | 699 | |||
XM_005262646.2 | XP_005262703.1 | 2 | 699 | |||
XM_005262647.2 | XP_005262704.1 | 2 | 699 | |||
XM_011513659.2 | XP_011511961.1 | 2 | 699 | |||
XM_011513660.2 | XP_011511962.1 | 2 | 699 | |||
XM_011513662.2 | XP_011511964.1 | 2 | 699 | |||
XM_011513663.2 | XP_011511965.1 | 2 | 699 | |||
XM_011513664.2 | XP_011511966.1 | 2 | 699 | |||
XM_011513665.2 | XP_011511967.1 | 2 | 699 | |||
XM_011513666.2 | XP_011511968.1 | 2 | 699 | |||
XM_011513668.2 | XP_011511970.1 | 2 | 699 | |||
XM_017007918.1 | XP_016863407.1 | 2 | 699 | |||
XM_017007919.1 | XP_016863408.1 | 2 | 699 | |||
XM_017007920.1 | XP_016863409.1 | 2 | 699 | |||
XM_017007922.1 | XP_016863411.1 | 2 | 699 | |||
GenBank mRNA | AB449881.1 | - | 1 | 341 | ||
AK295746.1 | - | 2 | 522 | |||
BC029950.1 | - | 2 | 643 | |||
BC050321.1 | - | 2 | 696 | 5262043 |