Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321729.1 | NP_001308658.1 | 16 | 1759 | ||
NM_001321730.1 | NP_001308659.1 | 16 | 1759 | |||
NM_001321731.1 | NP_001308660.1 | 16 | 1759 | |||
NM_001321733.1 | NP_001308662.1 | 16 | 1759 | |||
NM_001321734.1 | NP_001308663.1 | 15 | 1593 | |||
NM_015189.2 | NP_056004.1 | 16 | 1759 | |||
NR_135773.1 | 16 | 1759 | ||||
NR_135774.1 | 16 | 1759 | ||||
XM_005264224.1 | XP_005264281.1 | 9 | 1051 | |||
XM_011532711.2 | XP_011531013.1 | 16 | 1755 | |||
XM_011532712.2 | XP_011531014.1 | 16 | 1755 | |||
XM_017003641.1 | XP_016859130.1 | 12 | 1193 | |||
XM_017003642.1 | XP_016859131.1 | 10 | 806 | |||
GenBank mRNA | AB023136.1 | - | 16 | 1742 | ||
AK023791.1 | - | 4 | 320 | |||
AK026474.1 | - | 13 | 1254 |