Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIP5K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276277.1 | NP_001263206.1 | 26, 27 | 3698 | ||
NM_001281471.1 | NP_001268400.1 | 27, 28 | 3743 | |||
NM_015216.3 | NP_056031.2 | 25, 26 | 3463 | |||
XM_005271935.1 | XP_005271992.1 | 26, 27 | 3697 | |||
XM_005271936.1 | XP_005271993.1 | 25, 26 | 3523 | |||
XM_005277534.1 | XP_005277591.1 | 27, 28 | 3742 | |||
XM_005277535.1 | XP_005277592.1 | 27, 28 | 3739 | |||
XM_005277536.1 | XP_005277593.1 | 27, 28 | 3742 | |||
XM_005277538.1 | XP_005277595.1 | 26, 27 | 3568 | |||
XM_017009269.1 | XP_016864758.1 | 26, 27 | 3694 | |||
XM_017009272.1 | XP_016864761.1 | 25, 26 | 3523 | |||
XM_017009273.1 | XP_016864762.1 | 25, 26 | 3520 | |||
GenBank mRNA | AB007893.1 | - | 26, 27 | 3688 | ||
AK294787.1 | - | 27, 28 | 3607 | |||
BC024591.1 | - | 25, 26 | 3216 | 3913521 |