Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025010.4 | NP_079286.2 | 2 | 309 | ||
XM_005265001.3 | XP_005265058.2 | 2 | 209 | |||
XM_005265002.4 | XP_005265059.2 | 2 | 209 | |||
XM_005265003.3 | XP_005265060.2 | 2 | 209 | |||
XM_006713079.1 | XP_006713142.1 | 2 | 165 | |||
GenBank mRNA | AB062478.1 | - | 2 | 481 | ||
AI356423.1 | - | 2 | 309 | |||
AK023765.1 | - | 2 | 203 | |||
AK291477.1 | - | 2 | 203 | |||
BC032620.1 | - | 2 | 121 | 5578173 | ||
BX537953.1 | - | 2 | 200 |