Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025010.4 | NP_079286.2 | 4, 5 | 721 | ||
XM_005265001.3 | XP_005265058.2 | 4, 5 | 621 | |||
XM_005265002.4 | XP_005265059.2 | 4, 5 | 621 | |||
XM_005265003.3 | XP_005265060.2 | 4, 5 | 621 | |||
XM_006713079.1 | XP_006713142.1 | 4, 5 | 577 | |||
XM_017006033.1 | XP_016861522.1 | 3, 4 | 484 | |||
XM_017006034.1 | XP_016861523.1 | 3, 4 | 484 | |||
XM_017006035.1 | XP_016861524.1 | 3, 4 | 484 | |||
GenBank mRNA | AB018338.1 | - | 2, 3 | 269 | ||
AB062478.1 | - | 4, 5 | 893 | |||
AK023765.1 | - | 4, 5 | 616 | |||
AK291477.1 | - | 4, 5 | 615 | |||
AK295292.1 | - | 3, 4 | 483 | |||
AY714879.1 | - | 1, 2 | 148 | |||
BC032620.1 | - | 4, 5 | 533 | 5578173 | ||
BX537953.1 | - | 4, 5 | 612 |