Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286715.1 | NP_001273644.1 | 16 | 2502 | ||
NM_001286717.1 | NP_001273646.1 | 16 | 2382 | |||
NM_015239.2 | NP_056054.2 | 17 | 2447 | |||
XM_005251847.3 | XP_005251904.1 | 17 | 2513 | |||
XM_005251848.2 | XP_005251905.1 | 17 | 2691 | |||
XM_005251849.3 | XP_005251906.1 | 16 | 2437 | |||
XM_011518418.2 | XP_011516720.1 | 17 | 2513 | |||
XM_011518420.2 | XP_011516722.1 | 7 | 5766 | |||
XM_017014544.1 | XP_016870033.1 | 17 | 3058 | |||
XM_017014545.1 | XP_016870034.1 | 17 | 2393 | |||
XM_017014546.1 | XP_016870035.1 | 16 | 2280 | |||
XM_017014547.1 | XP_016870036.1 | 16 | 2437 | |||
XM_017014548.1 | XP_016870037.1 | 16 | 2380 | |||
XM_017014549.1 | XP_016870038.1 | 15 | 2399 | |||
GenBank mRNA | AB028958.2 | - | 17 | 2425 | ||
AK001544.1 | - | 18 | 2247 | |||
AK022562.1 | - | 5 | 1090 | |||
AK023280.1 | - | 5 | 1082 | |||
AK024698.1 | - | 1 | 32 | |||
AK295304.1 | - | 13 | 1941 | |||
AK295774.1 | - | 16 | 2382 | |||
AK299506.1 | - | 13 | 1942 | |||
AL833359.1 | - | 5 | 1039 | |||
BC060815.1 | - | 17 | 2412 | 30344122 | ||
BX648366.1 | - | 5 | 1494 |