Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142289.2 | NP_001135761.2 | 7 | 798 | ||
NM_001142290.2 | NP_001135762.1 | 7 | 798 | |||
NM_001142291.2 | NP_001135763.2 | 7 | 798 | |||
NM_015246.3 | NP_056061.1 | 7 | 798 | |||
NR_102267.1 | 7 | 798 | ||||
XM_005255217.1 | XP_005255274.1 | 7 | 801 | |||
XM_005255218.1 | XP_005255275.1 | 7 | 801 | |||
XM_005255219.3 | XP_005255276.1 | 7 | 801 | |||
XM_005255220.3 | XP_005255277.1 | 7 | 798 | |||
XM_005255221.3 | XP_005255278.1 | 7 | 798 | |||
XM_011522441.1 | XP_011520743.1 | 7 | 659 | |||
XM_017023091.1 | XP_016878580.1 | 7 | 801 | |||
XM_017023092.1 | XP_016878581.1 | 7 | 801 | |||
XM_017023093.1 | XP_016878582.1 | 7 | 656 | |||
GenBank mRNA | AB011116.1 | - | 7 | 754 | ||
AK299052.1 | - | 7 | 780 | |||
BC050389.1 | - | 7 | 770 | 5755971 | ||
EF471397.2 | - | 7 | 661 | |||
EF471398.2 | - | 7 | 661 |