Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015253.1 | NP_056068.1 | 7 | 1463 | ||
XM_005256572.1 | XP_005256629.1 | 7 | 1551 | |||
XM_005256573.1 | XP_005256630.1 | 7 | 1526 | |||
GenBank mRNA | AB011095.1 | - | 6 | 815 | ||
AK000243.1 | - | 5 | 936 | |||
AK289603.1 | - | 7 | 1522 | |||
AK309645.1 | - | 6 | 915 | |||
AL833730.1 | - | 7 | 1463 | |||
BC009975.2 | - | 7 | 1488 | 4110708 |